Learn More
Abcam ZK 756326, non-peptide CCR8 chemokine receptor agonist, 10MG
Catalog No. 502707234
Encompass_Preferred
MW 429.39 g/mol, Purity >99%. Non-peptide CCR8 chemokine receptor agonist (EC₅₀ = 245 nM). Demonstrates no agonist activity against CCR3, CXCR3, CXCR4 and CCR5. Desensitizes the receptor against CCL1 stimulation.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7234 Supplier Abcam Supplier No. AB12081410MG
MW 429.39 g/mol, Purity >99%. Non-peptide CCR8 chemokine receptor agonist (EC₅₀ = 245 nM). Demonstrates no agonist activity against CCR3, CXCR3, CXCR4 and CCR5. Desensitizes the receptor against CCL1 stimulation.
The product is subject to the following: Abcam Restricted Use Statement