Learn More
Cayman Chemical ZK 756326hydrochlorIde 10mg
SDP
Catalog No. 502938569
Encompass_Preferred
A non-peptide agonist of CCR8; inhibits binding of CCL1 to human CCR8 (IC50 = 1.8 µM); stimulates extracellular acidification in cells expressing human and mouse CCR8 (EC50s = 254 nM and 2.6 µM, respectively); exhibits >28-fold selectivity for CCR8 over 26 other GPCRs; binds to the serotonergic receptors 5-HT1A, 5-HT2B, 5-HT2C, and 5-HT5A (IC50s = 5.4, 4.4, 34.8, and 16 µM, respectively); induces Gαi-mediated calcium mobilization, ERK phosphorylation, exhibits leukocyte chemotactic activity, and inhibits HIV fusion of cells expressing CD4 and CCR8
Catalog No. 50-293-8569 Supplier Cayman Chemical Supplier No. 2845410 MG
A non-peptide agonist of CCR8; inhibits binding of CCL1 to human CCR8 (IC50 = 1.8 µM); stimulates extracellular acidification in cells expressing human and mouse CCR8 (EC50s = 254 nM and 2.6 µM, respectively); exhibits >28-fold selectivity for CCR8 over 26 other GPCRs; binds to the serotonergic receptors 5-HT1A, 5-HT2B, 5-HT2C, and 5-HT5A (IC50s = 5.4, 4.4, 34.8, and 16 µM, respectively); induces Gαi-mediated calcium mobilization, ERK phosphorylation, exhibits leukocyte chemotactic activity, and inhibits HIV fusion of cells expressing CD4 and CCR8