Learn More
TARGETMOL CHEMICALS INC ZK-90055 HYDROCHLORIDE 5MG
SDP
Catalog No. 502581349
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. ZK-90055 hydrochloride is a (beta)2 adrenergic receptor agonist used to study asthma. purity: 99%
Catalog No. 50-258-1349 Supplier TARGETMOL CHEMICALS INC Supplier No. T134035MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. ZK-90055 hydrochloride is a (beta)2 adrenergic receptor agonist used to study asthma. purity: 99%