Learn More
Medchemexpress LLC HY-12407 1mg , ZK168281 CAS:186371-96-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY124071MG
Encompass_Preferred
HY-12407 1mg ZK168281 CAS: 186371-96-0 ZK168281 is a 25-carboxylic ester 1α,25(OH)2D3 analog and a pure VDR antagonist with a Kd value of 0.1 nM. ZK168281 is an effective inhibitor of the coactivator (CoA) interaction of its receptor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1714