Learn More
Selleck Chemical LLC ZK756326 2HCl
SDP
Supplier:  Selleck Chemical LLC S83245MG
Encompass_Preferred
ZK756326 is a full agonist of CCR8 Chemokine receptor 8 with an IC50 of 1 8 M dose-responsively eliciting an increase in intracellular calcium and cross-desensitizing the response of the receptor to CCL1
Catalog No. 50-000-02888