Learn More
Medchemexpress LLC Zk756326 dihydrochloride | 1780259-94-0 | MFCD09038571 | 99.9% | 429.38 g/mol | C21H30Cl2N2O3 | 5 MG
SDP
Catalog No. 501932017
Encompass_Preferred
ZK756326 dihydrochloride is a selective, nonpeptide small-molecule agonist of the CC chemokine receptor CCR8 supplied as the dihydrochloride salt for research use. It is intended for in vitro pharmacology and receptor characterization studies and demonstrates potent CCR8 activity and high chemical purity.
  • Selective CCR8 agonist with IC50 1.8 μM in cellular assays.
  • High chemical purity suitable for biological assays and receptor profiling.
  • Dihydrochloride salt form offering improved handling and stability.
  • Good solubility in DMSO and aqueous buffers; ultrasonic assistance recommended.
  • Solid, white to off-white appearance for straightforward weighing and use.
  • Storage conditions compatible with short- and long-term sample preservation.
Catalog No. 50-193-2017 Supplier Medchemexpress LLC Supplier No. HY101038A5MG
ZK756326 dihydrochloride is a selective, nonpeptide small-molecule agonist of the CC chemokine receptor CCR8 supplied as the dihydrochloride salt for research use. It is intended for in vitro pharmacology and receptor characterization studies and demonstrates potent CCR8 activity and high chemical purity.
  • Selective CCR8 agonist with IC50 1.8 μM in cellular assays.
  • High chemical purity suitable for biological assays and receptor profiling.
  • Dihydrochloride salt form offering improved handling and stability.
  • Good solubility in DMSO and aqueous buffers; ultrasonic assistance recommended.
  • Solid, white to off-white appearance for straightforward weighing and use.
  • Storage conditions compatible with short- and long-term sample preservation.