Learn More
Medchemexpress LLC ZK756326 dihydrochloride | 1780259-94-0 | 99.9% | 429.38 | C21H30Cl2N2O3 | 10 MG
SDP
Catalog No. 5000212086
Encompass_Preferred
ZK756326 dihydrochloride is a small-molecule, nonpeptide agonist of the CC chemokine receptor CCR8 supplied for research use only. It is provided as a white to off-white solid with reported high purity and characterized molecular properties for pharmacology studies.
  • Nonpeptide agonist of the CC chemokine receptor CCR8.
  • High reported purity (≈99.9%).
  • Provided as a white to off-white solid.
  • Molecular weight 429.38 Da for analytical identification.
  • Supplied in milligram quantities for laboratory research.
Catalog No. 50-002-12086 Supplier Medchemexpress LLC Supplier No. HY101038A10MG
ZK756326 dihydrochloride is a small-molecule, nonpeptide agonist of the CC chemokine receptor CCR8 supplied for research use only. It is provided as a white to off-white solid with reported high purity and characterized molecular properties for pharmacology studies.
  • Nonpeptide agonist of the CC chemokine receptor CCR8.
  • High reported purity (≈99.9%).
  • Provided as a white to off-white solid.
  • Molecular weight 429.38 Da for analytical identification.
  • Supplied in milligram quantities for laboratory research.