Learn More
Medchemexpress LLC ZK756326 dihydrochloride | 1780259-94-0 | 99.9% | 429.38 g/mol | C21H30Cl2N2O3 | 100 MG
SDP
Catalog No. 5000213593
Encompass_Preferred
ZK756326 dihydrochloride is a selective, non-peptide agonist of the CC chemokine receptor CCR8 supplied as the dihydrochloride salt for research use. The compound is provided as a high-purity solid intended for pharmacology and receptor biology studies, including in vitro receptor activation and mechanism-of-action experiments.
  • High purity suitable for research assays.
  • Selective agonist activity at CCR8 for receptor pharmacology.
  • Supplied as a stable dihydrochloride salt for handling and formulation.
  • Appropriate for in vitro signaling and binding studies.
  • Store sealed and away from moisture to preserve stability.
Catalog No. 50-002-13593 Supplier Medchemexpress LLC Supplier No. HY101038A100MG
ZK756326 dihydrochloride is a selective, non-peptide agonist of the CC chemokine receptor CCR8 supplied as the dihydrochloride salt for research use. The compound is provided as a high-purity solid intended for pharmacology and receptor biology studies, including in vitro receptor activation and mechanism-of-action experiments.
  • High purity suitable for research assays.
  • Selective agonist activity at CCR8 for receptor pharmacology.
  • Supplied as a stable dihydrochloride salt for handling and formulation.
  • Appropriate for in vitro signaling and binding studies.
  • Store sealed and away from moisture to preserve stability.