Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zk756326 dihydrochloride | 1780259-94-0 | MFCD09038571 | 99.9% | 429.38 | C21H30Cl2N2O3 | 1 ML
SDP

Catalog No. 5000211174
Encompass_Preferred

ZK756326 dihydrochloride is a small-molecule, nonpeptide agonist of the CC chemokine receptor CCR8 supplied for research use in biochemical and cell-based assays. It is available as a 10 mM solution in DMSO (1 mL) and as crystalline solids, and is provided with analytical documentation to verify identity and purity.

  • Nonpeptide CCR8 agonist suitable for receptor pharmacology studies.
  • Offered as 10 mM solution in DMSO (1 mL) and in multiple solid quantities for assay flexibility.
  • High purity (99.87%) supporting reproducible experimental results.
  • Comes with analytical documentation: COA, HNMR, LCMS, and SDS.
  • Storage guidance for stability: sealed storage; in solvent, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-11174 Supplier Medchemexpress LLC Supplier No. HY101038A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZK756326 dihydrochloride is a small-molecule, nonpeptide agonist of the CC chemokine receptor CCR8 supplied for research use in biochemical and cell-based assays. It is available as a 10 mM solution in DMSO (1 mL) and as crystalline solids, and is provided with analytical documentation to verify identity and purity.

  • Nonpeptide CCR8 agonist suitable for receptor pharmacology studies.
  • Offered as 10 mM solution in DMSO (1 mL) and in multiple solid quantities for assay flexibility.
  • High purity (99.87%) supporting reproducible experimental results.
  • Comes with analytical documentation: COA, HNMR, LCMS, and SDS.
  • Storage guidance for stability: sealed storage; in solvent, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.