Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ZK756326 dihydrochloride | 1780259-94-0 | MFCD09038571 | 99.9% | 429.38 | C21H30Cl2N2O3 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY101038A50MG

Encompass_Preferred

ZK756326 dihydrochloride is a nonpeptide agonist of the CC chemokine receptor CCR8 used as a research tool to study CCR8 signaling. It inhibits binding of the CCR8 ligand I-309 (CCL1) with an IC50 of 1.8 μM, functions as a full agonist that increases intracellular calcium, and stimulates extracellular acidification in CCR8-expressing cells.

  • Nonpeptide CCR8 agonist for receptor activation studies.
  • IC50 for CCR8 binding inhibition: 1.8 μM.
  • Full agonist - induces dose-dependent intracellular calcium increase.
  • Shows >28-fold specificity versus a 26-GPCR panel at 50 μM.
  • Reduced selectivity observed versus certain serotonergic and α2A-adrenergic receptors.
  • Purity 99.87%, white to off-white solid.
  • Available as multiple solid sizes and 10 mM solution in DMSO.
  • Storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-12824


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.