Learn More
Medchemexpress LLC ZK756326 dihydrochloride | 1780259-94-0 | MFCD09038571 | 99.9% | 429.38 g/mol | C21H30Cl2N2O3 | 25 MG
SDP
Catalog No. 5000212823
Encompass_Preferred
ZK756326 dihydrochloride is a small-molecule, nonpeptide agonist of the CC chemokine receptor CCR8 supplied as the dihydrochloride salt for research use only. It is intended for in vitro pharmacology and preclinical studies where modulation of CCR8 signaling is required, and is provided with a defined formula, molecular weight, and high purity.
  • Selective CCR8 agonist for receptor pharmacology studies.
  • High purity suitable for analytical and biological assays (99.87%).
  • Well-defined chemical identity with molecular formula and weight.
  • Available in small pack sizes for laboratory research.
  • For research use only; not for human or veterinary applications.
Catalog No. 50-002-12823 Supplier Medchemexpress LLC Supplier No. HY101038A25MG
ZK756326 dihydrochloride is a small-molecule, nonpeptide agonist of the CC chemokine receptor CCR8 supplied as the dihydrochloride salt for research use only. It is intended for in vitro pharmacology and preclinical studies where modulation of CCR8 signaling is required, and is provided with a defined formula, molecular weight, and high purity.
  • Selective CCR8 agonist for receptor pharmacology studies.
  • High purity suitable for analytical and biological assays (99.87%).
  • Well-defined chemical identity with molecular formula and weight.
  • Available in small pack sizes for laboratory research.
  • For research use only; not for human or veterinary applications.