Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ZK824190 hydrochloride | 2629177-12-2 | 99.2% | 450.86 g/mol | C22H21ClF2N2O4 | 10MM 1ML
SDP

Catalog No. 5000238776
Encompass_Preferred

ZK824190 hydrochloride is the hydrochloride salt of a selective, orally available urokinase plasminogen activator (uPA) inhibitor used in research into uPA biology and potential therapies for multiple sclerosis. It is supplied as a high-purity solid and as a pre-made 10 mM solution in DMSO for in vitro and preclinical studies; not for human use.

  • Selective urokinase plasminogen activator (uPA) inhibitor.
  • Supplied as 10 mM solution in DMSO and as solid quantities.
  • High purity suitable for research applications.
  • Molecular formula: C22H21ClF2N2O4.
  • Molecular weight: 450.86 g/mol.
  • CAS: 2629177-12-2 for substance identification.
  • Provided as the hydrochloride salt for stability and handling.
  • Intended for in vitro and preclinical research only, not for human use.

Catalog No. 50-002-38776 Supplier Medchemexpress LLC Supplier No. HY126361A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZK824190 hydrochloride is the hydrochloride salt of a selective, orally available urokinase plasminogen activator (uPA) inhibitor used in research into uPA biology and potential therapies for multiple sclerosis. It is supplied as a high-purity solid and as a pre-made 10 mM solution in DMSO for in vitro and preclinical studies; not for human use.

  • Selective urokinase plasminogen activator (uPA) inhibitor.
  • Supplied as 10 mM solution in DMSO and as solid quantities.
  • High purity suitable for research applications.
  • Molecular formula: C22H21ClF2N2O4.
  • Molecular weight: 450.86 g/mol.
  • CAS: 2629177-12-2 for substance identification.
  • Provided as the hydrochloride salt for stability and handling.
  • Intended for in vitro and preclinical research only, not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.