Learn More
Medchemexpress LLC HY-114330A 10mg Medchemexpress, ZK824859 hydrochloride CAS: Purity:>98%
SDP
Catalog No. 501969328
Encompass_Preferred
Medchemexpress, HY-114330A 10mg ZK824859 HCl CAS: ZK824859 hydrochloride is an oral available and selective urokinase plasminogen activator (uPA) inhibitor with IC50s of 79 nM, 1580 nM and 1330 nM for human uPA, tPA, and plasmin, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9328 Supplier Medchemexpress LLC Supplier No. HY114330A10MG
Medchemexpress, HY-114330A 10mg ZK824859 HCl CAS: ZK824859 hydrochloride is an oral available and selective urokinase plasminogen activator (uPA) inhibitor with IC50s of 79 nM, 1580 nM and 1330 nM for human uPA, tPA, and plasmin, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.