Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ZK824859 hydrochlori 50mg | 2436760-76-6 | 50 MG
SDP

Catalog No. 5000229766
Encompass_Preferred

ZK824859 hydrochloride is a research-stage small-molecule hydrochloride salt that functions as an orally available, selective inhibitor of urokinase plasminogen activator (uPA). Reported potency includes IC50 values of 79 nM for human uPA, 1,580 nM for tPA, and 1,330 nM for plasmin. The compound is supplied as a solid powder for in vitro and in vivo research use.

  • Orally available uPA inhibitor.
  • Selective activity: human uPA IC50 = 79 nM; tPA = 1,580 nM; plasmin = 1,330 nM.
  • Hydrochloride salt form for improved solubility.
  • CAS 2436760-76-6; molecular formula C23H23ClF2N2O4; molecular weight 464.89 g·mol-1.
  • Common research package sizes include 25 mg and 50 mg.

Catalog No. 50-002-29766 Supplier Medchemexpress LLC Supplier No. HY114330A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZK824859 hydrochloride is a research-stage small-molecule hydrochloride salt that functions as an orally available, selective inhibitor of urokinase plasminogen activator (uPA). Reported potency includes IC50 values of 79 nM for human uPA, 1,580 nM for tPA, and 1,330 nM for plasmin. The compound is supplied as a solid powder for in vitro and in vivo research use.

  • Orally available uPA inhibitor.
  • Selective activity: human uPA IC50 = 79 nM; tPA = 1,580 nM; plasmin = 1,330 nM.
  • Hydrochloride salt form for improved solubility.
  • CAS 2436760-76-6; molecular formula C23H23ClF2N2O4; molecular weight 464.89 g·mol-1.
  • Common research package sizes include 25 mg and 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.