Learn More
Aviva Systems Biology Corp ZKSCAN1 Antibody - middle region (ARP47580_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47580_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZKSCAN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZKSCAN1. Alternate names: 9130423L19Rik, KOX18, MGC138429, PHZ-37, ZNF139, ZNF36ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59597