Learn More
Aviva Systems Biology Corp Zkscan14 Antibody - middle region (ARP37322_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37322_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zkscan14. It was validated on Western Blot. Zkscan14,zinc finger with KRAB and SCAN domains 14,Zfp99; Znf394; MGC117641; 2310046C23Rik; 2810437E14Rik; Zkscan14,zinc finger protein 394,zinc finger protein 394,zfp-94,OTTMUSP00000026365,zinc finger protein 94,zinc finger protein 99,zinc finger protein with KRAB and SCAN domains 14The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2310046C23Rik, 2810437E14Rik, MGC117641, Zfp99, Znf394ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-615