Learn More
Aviva Systems Biology Corp Zkscan14 Antibody - C-terminal region (ARP39683_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39683_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Zkscan14. It was validated on Western Blot. . Alternate names: 2310046C23Rik, 2810437E14Rik, MGC117641, Zfp99, Znf394ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-821