Learn More
Aviva Systems Biology Corp ZKSCAN5 Antibody - N-terminal region (ARP32522_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32522_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZKSCAN5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZKSCAN5. Alternate names: FLJ39233, KIAA1015, MGC33710, ZFP95, ZNF914ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-54634