Learn More
Medchemexpress LLC HY-100456 5mg Medchemexpress, ZL006 CAS:1181226-02-7 Purity:>98%
SDP
Catalog No. 501931842
Encompass_Preferred
Medchemexpress, HY-100456 5mg ZL006 CAS:1181226-02-7 ZL006 is a potent inhibitor of nNOS/PSD-95 interaction, and inhibits NMDA receptor -mediated NO synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1842 Supplier Medchemexpress LLC Supplier No. HY1004565MG
Medchemexpress, HY-100456 5mg ZL006 CAS:1181226-02-7 ZL006 is a potent inhibitor of nNOS/PSD-95 interaction, and inhibits NMDA receptor -mediated NO synthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.