Learn More
Medchemexpress LLC HY-112149 10mg Medchemexpress, ZL0420 CAS:2230496-80-5 Purity:>98%
SDP
Catalog No. 501969039
Encompass_Preferred
Medchemexpress, HY-112149 10mg ZL0420 CAS:2230496-80-5 ZL0420 is a potent and selective bromodomain-containing protein 4 (BRD4) inhibitor with IC50 values of 27 nM against BRD4 BD1 and 32 nM against BRD4 BD2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9039 Supplier Medchemexpress LLC Supplier No. HY11214910MG
Medchemexpress, HY-112149 10mg ZL0420 CAS:2230496-80-5 ZL0420 is a potent and selective bromodomain-containing protein 4 (BRD4) inhibitor with IC50 values of 27 nM against BRD4 BD1 and 32 nM against BRD4 BD2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.