Learn More
Medchemexpress LLC Zl0454 | 2229042-77-5 | 95.0% | 374.46 | 100 MG
SDP
Catalog No. 5000437445
Encompass_Preferred
ZL0454 is a potent and selective Bromodomain-containing protein 4 (BRD4) inhibitor with an IC50 of 49 and 32 nM for BD1 and BD2.
  • Potent and selective bromodomain-containing protein 4 (BRD4) inhibitor.
  • IC50 values of 49 nM for BD1 and 32 nM for BD2.
Catalog No. 50-004-37445 Supplier Medchemexpress LLC Supplier No. HY112150100MG
ZL0454 is a potent and selective Bromodomain-containing protein 4 (BRD4) inhibitor with an IC50 of 49 and 32 nM for BD1 and BD2.
  • Potent and selective bromodomain-containing protein 4 (BRD4) inhibitor.
  • IC50 values of 49 nM for BD1 and 32 nM for BD2.