Learn More
Medchemexpress LLC ZL0580 | 2377151-10-3 | 99.4% | 532.53 | C25H23F3N4O4S | 5 MG
SDP
Catalog No. 5000264890
Encompass_Preferred
ZL0580 is a BRD4 BD1-binding small molecule that induces epigenetic suppression of HIV by inhibiting Tat transactivation and transcription elongation and by promoting a repressive chromatin structure at the HIV promoter. It is supplied as a high-purity solid or as DMSO solutions for research applications and is stable under recommended storage conditions.
Catalog No. 50-002-64890 Supplier Medchemexpress LLC Supplier No. HY1264285MG
ZL0580 is a BRD4 BD1-binding small molecule that induces epigenetic suppression of HIV by inhibiting Tat transactivation and transcription elongation and by promoting a repressive chromatin structure at the HIV promoter. It is supplied as a high-purity solid or as DMSO solutions for research applications and is stable under recommended storage conditions.