Learn More
Medchemexpress LLC Zl0590 | 2230496-99-6 | MFCD34604141 | 99.9% | C23H27F3N4O4S | 10MG
SDP
Catalog No. 5000261436
Encompass_Preferred
ZL0590 is a potent, orally active BRD4 BD1-selective inhibitor (IC50 = 90 nM) used for research into BRD4-mediated biology and inflammatory disease models. It has demonstrated anti-inflammatory effects in animal models and is provided in both solid form and as a DMSO solution, with high reported purity for research use.
  • Potent BRD4 BD1 inhibition with low-nanomolar activity.
  • Demonstrated anti-inflammatory activity in animal models.
  • Suitable for in vitro and in vivo research applications.
  • Available as solid and 10 mM solution in DMSO for flexibility.
  • High reported purity for research-grade experiments.
Catalog No. 50-002-61436 Supplier Medchemexpress LLC Supplier No. HY14531010MG
ZL0590 is a potent, orally active BRD4 BD1-selective inhibitor (IC50 = 90 nM) used for research into BRD4-mediated biology and inflammatory disease models. It has demonstrated anti-inflammatory effects in animal models and is provided in both solid form and as a DMSO solution, with high reported purity for research use.
  • Potent BRD4 BD1 inhibition with low-nanomolar activity.
  • Demonstrated anti-inflammatory activity in animal models.
  • Suitable for in vitro and in vivo research applications.
  • Available as solid and 10 mM solution in DMSO for flexibility.
  • High reported purity for research-grade experiments.