Learn More
TARGETMOL CHEMICALS INC ZL0590 5MG
SDP
Catalog No. 502579683
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ZL0590 is an effective and selective inhibitor of BD1-BRD4 (IC50 = 90 nM) with anti-inflammatory activities. purity: 99%
Catalog No. 50-257-9683 Supplier TARGETMOL CHEMICALS INC Supplier No. T60072
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ZL0590 is an effective and selective inhibitor of BD1-BRD4 (IC50 = 90 nM) with anti-inflammatory activities. purity: 99%