Learn More
Medchemexpress LLC HY-17538 10mg , ZLN005 CAS:49671-76-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1753810MG
Encompass_Preferred
HY-17538 10mg ZLN005 CAS: 49671-76-3 ZLN005 is a potent activator of peroxisome proliferator-activated receptor-γ coactivator-1α (PGC-1α). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9154