Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ZLN005-d4 | 2410443-42-2 | 99.5% | 254.36 g·mol⁻1 | C17H14D4N2 | 10 MG
SDP

Catalog No. 5000299210
Encompass_Preferred

ZLN005-d4 is the deuterium-labeled analogue of ZLN005, a small-molecule activator of PGC-1α (peroxisome proliferator-activated receptor-γ coactivator-1α). Supplied as a white to off-white solid with high analytical purity, it is intended for use as a tracer and as an internal standard in quantitative NMR, GC-MS, and LC-MS workflows.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity (99.5%) suitable for analytical and tracer applications.
  • Molecular formula C17H14D4N2; molecular weight 254.36 g·mol⁻1.
  • Solid, white to off-white appearance for straightforward handling.
  • Recommended storage: powder -20 °C (long term) or 4 °C (intermediate); in solvent -80 °C (short term).
  • Useful as a tracer or internal standard for NMR, GC-MS, and LC-MS quantitation.

Catalog No. 50-002-99210 Supplier Medchemexpress LLC Supplier No. HY17538S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZLN005-d4 is the deuterium-labeled analogue of ZLN005, a small-molecule activator of PGC-1α (peroxisome proliferator-activated receptor-γ coactivator-1α). Supplied as a white to off-white solid with high analytical purity, it is intended for use as a tracer and as an internal standard in quantitative NMR, GC-MS, and LC-MS workflows.

  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity (99.5%) suitable for analytical and tracer applications.
  • Molecular formula C17H14D4N2; molecular weight 254.36 g·mol⁻1.
  • Solid, white to off-white appearance for straightforward handling.
  • Recommended storage: powder -20 °C (long term) or 4 °C (intermediate); in solvent -80 °C (short term).
  • Useful as a tracer or internal standard for NMR, GC-MS, and LC-MS quantitation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.