Learn More
Abcam ZLN005, PGC-1alpha transcriptional activator, 10MG
Catalog No. 502705086
Encompass_Preferred
MW 250.34 Da, Purity >95%. PGC-1α transcriptional activator. Activates AMPK. Stimulates PGC-1α expression. Increases fat oxidation. Improves glucose and pyruvate tolerance. Increases insulin sensitivity. Shows antidiabetic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5086 Supplier Abcam Supplier No. AB14564110MG
MW 250.34 Da, Purity >95%. PGC-1α transcriptional activator. Activates AMPK. Stimulates PGC-1α expression. Increases fat oxidation. Improves glucose and pyruvate tolerance. Increases insulin sensitivity. Shows antidiabetic effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement