Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ZLN024 hydrochloride | 1883548-91-1 | 98.5% | 361.69 | 100 MG
SDP

Catalog No. 5000393530
Encompass_Preferred

ZLN024 hydrochloride is an allosteric activator of AMPK. It directly activates recombinant AMPK α1β1γ1, AMPK α2β1γ1, AMPK α1β2γ1, and AMPK α2β2γ1 heterotrimers.

  • Allosterically stimulates active AMPK heterotrimers and inactive α1 subunit truncations.
  • AMPK activation requires pre-phosphorylation of Thr-172 and protects AMPK Thr-172 against dephosphorylation by PP2Cα.
  • Activates AMPK in L6 myotubes, stimulating glucose uptake and fatty acid oxidation without increasing the ADP/ATP ratio.
  • Increases the activity of α1β1γ1 by 1.5-fold (EC50 of 0.42 μM), α2β1γ1 by 1.7-fold (EC50 of 0.95 μM), α1β2γ1 by 1.7-fold (EC50 of 1.1 μM), and α2β2γ1 by 1.6-fold (EC50 of 0.13 μM).
  • Improves glucose tolerance in C57BKS db/db mice after 4 weeks of treatment.
  • Reduces fasting blood glucose by 15%, liver tissue weight, triacylglycerol, and total cholesterol content in mice.

Catalog No. 50-003-93530 Supplier Medchemexpress LLC Supplier No. HY16708A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZLN024 hydrochloride is an allosteric activator of AMPK. It directly activates recombinant AMPK α1β1γ1, AMPK α2β1γ1, AMPK α1β2γ1, and AMPK α2β2γ1 heterotrimers.

  • Allosterically stimulates active AMPK heterotrimers and inactive α1 subunit truncations.
  • AMPK activation requires pre-phosphorylation of Thr-172 and protects AMPK Thr-172 against dephosphorylation by PP2Cα.
  • Activates AMPK in L6 myotubes, stimulating glucose uptake and fatty acid oxidation without increasing the ADP/ATP ratio.
  • Increases the activity of α1β1γ1 by 1.5-fold (EC50 of 0.42 μM), α2β1γ1 by 1.7-fold (EC50 of 0.95 μM), α1β2γ1 by 1.7-fold (EC50 of 1.1 μM), and α2β2γ1 by 1.6-fold (EC50 of 0.13 μM).
  • Improves glucose tolerance in C57BKS db/db mice after 4 weeks of treatment.
  • Reduces fasting blood glucose by 15%, liver tissue weight, triacylglycerol, and total cholesterol content in mice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.