Learn More
Cayman Chemical Z-LRGG-AMC (trifluoroacetate salt), Purity: ≥95%, 50 mg
SDP
Catalog No. 502933855
Encompass_Preferred
A fluorogenic substrate for isopeptidase T; AMC is released upon enzymatic cleavage by isopeptidase T and its fluorescence can be used to quantify isopeptidase T activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-293-3855 Supplier Cayman Chemical Supplier No. 2664150 MG
A fluorogenic substrate for isopeptidase T; AMC is released upon enzymatic cleavage by isopeptidase T and its fluorescence can be used to quantify isopeptidase T activity; ex/em = 340-360/440-460 nm, respectively