Learn More
CPC Scientific Z-Leu-Val-Gly-DMK 0.5MG
SDP
Supplier:  CPC Scientific SUBS042A
Encompass
SEQUENCE: Z-Leu-Val-Gly-DMKONE-LETTER SEQUENCE: ZLVG-DMKMOLECULAR FORMULA: C22H31N5O5MOLECULAR WEIGHT: 445.52STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [119670-30-3]SYNONYMS: Z-Leu-Val-Gly-diazomethylketoneRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:SKU(s): SUBS-042A, SUBS-042B, SUBS-042C, SUBS-042D
Catalog No. 50-211-0524