Learn More
TARGETMOL CHEMICALS INC ZM 226600 5MG
SDP
Catalog No. 502580856
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ZM 226600 is an ATP-sensitive potassium channel opener with an EC50 value of 500 nM. ZM226600 has an inhibitory effect on spontaneous bladder activity. purity: 100%
Catalog No. 50-258-0856 Supplier TARGETMOL CHEMICALS INC Supplier No. T235635MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. ZM 226600 is an ATP-sensitive potassium channel opener with an EC50 value of 500 nM. ZM226600 has an inhibitory effect on spontaneous bladder activity. purity: 100%