Learn More
Apexbio Technology LLC ZM 241385 139180-30-6 200mg
SDP
Catalog No. 5000091910
Encompass_Preferred
ZM 241385 is a selective high-affinity antagonist targeting the adenosine A2A receptor (A2AR) It is designed to inhibit A2AR-mediated signaling thereby modulating receptor-specific downstream pathways ZM 241385 exerts its biological activity primarily through competitive inhibition of A2AR It demonstrates antagonistic activity with a Ki value of approximately 1 4 nM Based on these pharmacological properties ZM 241385 holds research potential in studies of neurodegenerative disorders cardiovascular conditions and A2AR-mediated neuronal signaling pathways
Catalog No. 50-000-91910 Supplier Apexbio Technology LLC Supplier No. B6561200
ZM 241385 is a selective high-affinity antagonist targeting the adenosine A2A receptor (A2AR) It is designed to inhibit A2AR-mediated signaling thereby modulating receptor-specific downstream pathways ZM 241385 exerts its biological activity primarily through competitive inhibition of A2AR It demonstrates antagonistic activity with a Ki value of approximately 1 4 nM Based on these pharmacological properties ZM 241385 holds research potential in studies of neurodegenerative disorders cardiovascular conditions and A2AR-mediated neuronal signaling pathways