Learn More
Enzo Life Sciences ZM-306416 (1mg). CAS: 690206-97-4
SDP
Supplier:  Enzo Life Sciences BMLEI3530001
Encompass_Preferred
VEGF receptor tyrosine kinase inhibitor. Inhibits KDR (IC50=100nM) and Flt (IC50=2µM) tyrosine kinases. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (3 mg/ml) or ethanol (1 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0739