Learn More
Enzo Life Sciences ZM-323881 (1mg). CAS: 193001-14-8
SDP
Supplier:  Enzo Life Sciences BMLEI3850001
Encompass_Preferred
Potent and selective inhibitor of the kinase activity of the human vascular endothelial growth factor receptor 2 (VEGFR2/KDR). Selectively inhibits VEGFR2 over VEGFR1 and other receptor tyrosine kinases with an IC50 = 2nM. Useful tool for exploring the biology of VEGFR receptors. Alternative name: 5-((7-Benzyloxyquinazolin-4-yl)amino)-4-fluoro-2-methyl phenol. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (3 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0762