Learn More
Selleck Chemical LLC ZM 39923 HCl 10mg 1021868-92-7
SDP
Catalog No. 501365535
Encompass_Preferred
ZM 39923 HCl is an JAK1/3 inhibitor with pIC50 of 4.4/7.1, almost no activity to JAK2 and modestly potent to EGFR; also found to be sensitive to transglutaminase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5535 Supplier Selleck Chemical LLC Supplier No. S800410MG
ZM 39923 HCl is an JAK1/3 inhibitor with pIC50 of 4.4/7.1, almost no activity to JAK2 and modestly potent to EGFR; also found to be sensitive to transglutaminase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.