Learn More
Apexbio Technology LLC ZM 447439 331771-20-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061537
Encompass_Preferred
ZM 447439 (CAS 331771-20-1) is a selective inhibitor of Aurora kinases a subgroup within the serine/threonine kinase family involved in chromosome segregation during cell division It inhibits recombinant Aurora A and Aurora B kinases with IC50 values of approximately 110 nM and 130 nM respectively consequently reducing phosphorylation of histone H3 at serine 10 ZM 447439 demonstrates limited inhibitory activity towards other kinases with IC50 values above 10 M for CDK1/2/4 IKK1/2 PLK1 CHK1 cFLT2 KDR2 FAK and Zap-70 In biological research ZM 447439 is commonly employed to study mitotic regulation chromosomal alignment and apoptosis signaling pathways
Catalog No. 50-000-61537 Supplier Apexbio Technology LLC Supplier No. A41135.1
ZM 447439 (CAS 331771-20-1) is a selective inhibitor of Aurora kinases a subgroup within the serine/threonine kinase family involved in chromosome segregation during cell division It inhibits recombinant Aurora A and Aurora B kinases with IC50 values of approximately 110 nM and 130 nM respectively consequently reducing phosphorylation of histone H3 at serine 10 ZM 447439 demonstrates limited inhibitory activity towards other kinases with IC50 values above 10 M for CDK1/2/4 IKK1/2 PLK1 CHK1 cFLT2 KDR2 FAK and Zap-70 In biological research ZM 447439 is commonly employed to study mitotic regulation chromosomal alignment and apoptosis signaling pathways