Learn More
Selleck Chemical LLC ZM 447439 25mg 331771-20-1
SDP
Catalog No. 508280
Encompass_Preferred
ZM 447439 is a selective and ATP-competitive inhibitor for Aurora A and Aurora B with IC50 of 110 nM and 130 nM, respectively. It is more than 8-fold selective for Aurora A/B than MEK1, Src, Lck and has little effect against CDK1/2/4, Plk1, Chk1, etc. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-828-0 Supplier Selleck Chemical LLC Supplier No. S110325MG
ZM 447439 is a selective and ATP-competitive inhibitor for Aurora A and Aurora B with IC50 of 110 nM and 130 nM, respectively. It is more than 8-fold selective for Aurora A/B than MEK1, Src, Lck and has little effect against CDK1/2/4, Plk1, Chk1, etc. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.