Learn More
Medchemexpress LLC HY-101790A 100mg Medchemexpress, ZM223 (hydrochloride) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101790A100MG
Encompass_Preferred
Medchemexpress, HY-101790A 100mg ZM223 (hydrochloride) CAS: ZM223 hydrochloride is an orally active, potent non-covalent NEDD8 activating enzyme ( NAE ) inhibitor with excellent anticancer activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2217