Learn More
Medchemexpress LLC Zm223 hydrochloride | 2438679-27-5 | 98.9% | 538.99 g/mol | C23H18ClF3N4O2S2 | 10 MG
SDP
Catalog No. 5000248367
Encompass_Preferred
ZM223 hydrochloride is a small-molecule, orally active, non-covalent inhibitor of the NEDD8-activating enzyme (NAE) with reported anticancer activity. Supplied as the hydrochloride salt, it is used in biochemical and cellular studies to probe the neddylation pathway and evaluate antiproliferative effects.
  • Orally active non-covalent NAE inhibitor.
  • Reported anticancer activity in cellular models.
  • Suitable for biochemical and cellular assays.
  • Provided as the hydrochloride salt for improved handling.
  • High purity (≈98.9%) suitable for research use.
Catalog No. 50-002-48367 Supplier Medchemexpress LLC Supplier No. HY101790A10MG
ZM223 hydrochloride is a small-molecule, orally active, non-covalent inhibitor of the NEDD8-activating enzyme (NAE) with reported anticancer activity. Supplied as the hydrochloride salt, it is used in biochemical and cellular studies to probe the neddylation pathway and evaluate antiproliferative effects.
  • Orally active non-covalent NAE inhibitor.
  • Reported anticancer activity in cellular models.
  • Suitable for biochemical and cellular assays.
  • Provided as the hydrochloride salt for improved handling.
  • High purity (≈98.9%) suitable for research use.