Learn More
Selleck Chemical LLC ZM241385-10mg
SDP
Catalog No. 502390069
Encompass_Preferred
ZM-241385 is a high affinity antagonist ligand selective for the adenosine A2A receptor.
Catalog No. 50-239-0069 Supplier Selleck Chemical LLC Supplier No. S810510MG
ZM-241385 is a high affinity antagonist ligand selective for the adenosine A2A receptor.