Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ZM323881 hydrochloride | 193000-39-4 | C22H19ClFN3O2 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY15467A10MM/1ML

Encompass_Preferred

ZM323881 hydrochloride is a potent and selective inhibitor of Vascular Endothelial Growth Factor Receptor 2 (VEGFR2) with an IC50 of less than 2 nM. This anilinoquinazoline specifically targets VEGFR2 tyrosine kinase activity, demonstrating excellent selectivity over other receptor tyrosine kinases.

  • Potent and selective VEGFR2 inhibitor
  • High purity (99.63%)
  • Specifically targets VEGFR2 tyrosine kinase activity
  • Exhibits excellent selectivity over other receptor tyrosine kinases
  • Inhibits VEGF-A-induced endothelial cell proliferation
  • Inhibits VEGFR2 tyrosine phosphorylation
  • Molecular Weight: 411.86
  • Formula: C22H19ClFN3O2
  • Appearance: Solid, light yellow to yellow

Catalog No. 50-003-76493


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.