Learn More
Medchemexpress LLC Zm323881 hydrochloride | 193000-39-4 | C22H19ClFN3O2 | 25 MG
SDP
Catalog No. 5000374115
Encompass_Preferred
ZM323881 hydrochloride is a potent and selective VEGFR2 inhibitor with an IC50 of less than 2 nM. This anilinoquinazoline potently inhibits VEGFR2 (KDR) tyrosine kinase activity, showing excellent selectivity over other receptor tyrosine kinases. It inhibits VEGF-A-induced endothelial cell proliferation and VEGFR2 tyrosine phosphorylation.
  • Potent and selective VEGFR2 inhibitor (IC50 < 2 nM)
  • Inhibits VEGFR2 (KDR) tyrosine kinase activity
  • Excellent selectivity over other receptor tyrosine kinases
  • Inhibits VEGF-A-induced endothelial cell proliferation (IC50 = 8 nM)
  • Inhibits VEGFR2 tyrosine phosphorylation
  • Blocks activation of VEGFR-2, while sparing VEGFR-1, EGFR, PDGFR, and HGF receptor
  • For research use only
Catalog No. 50-003-74115 Supplier Medchemexpress LLC Supplier No. 25MGHY15467A
ZM323881 hydrochloride is a potent and selective VEGFR2 inhibitor with an IC50 of less than 2 nM. This anilinoquinazoline potently inhibits VEGFR2 (KDR) tyrosine kinase activity, showing excellent selectivity over other receptor tyrosine kinases. It inhibits VEGF-A-induced endothelial cell proliferation and VEGFR2 tyrosine phosphorylation.
  • Potent and selective VEGFR2 inhibitor (IC50 < 2 nM)
  • Inhibits VEGFR2 (KDR) tyrosine kinase activity
  • Excellent selectivity over other receptor tyrosine kinases
  • Inhibits VEGF-A-induced endothelial cell proliferation (IC50 = 8 nM)
  • Inhibits VEGFR2 tyrosine phosphorylation
  • Blocks activation of VEGFR-2, while sparing VEGFR-1, EGFR, PDGFR, and HGF receptor
  • For research use only