Learn More
Medchemexpress LLC Zm323881 (hydrochloride) | 193000-39-4 | C22H19ClFN3O2 | 50 MG
SDP
Catalog No. 5000376212
Encompass_Preferred
ZM323881 hydrochloride is a potent and selective VEGFR2 inhibitor with an IC50 of less than 2 nM. It is an anilinoquinazoline that potently inhibits VEGFR2 (KDR) tyrosine kinase activity, demonstrating excellent selectivity versus other receptor tyrosine kinases. It inhibits VEGF-A-induced endothelial cell proliferation (IC50=8 nM) and VEGFR2 tyrosine phosphorylation, making it suitable for research applications.
  • Potent and selective VEGFR2 inhibitor
  • Anilinoquinazoline structure
  • Inhibits VEGF-A-induced endothelial cell proliferation
  • Inhibits VEGFR2 tyrosine phosphorylation
  • For research use only
Catalog No. 50-003-76212 Supplier Medchemexpress LLC Supplier No. HY15467A50MG
ZM323881 hydrochloride is a potent and selective VEGFR2 inhibitor with an IC50 of less than 2 nM. It is an anilinoquinazoline that potently inhibits VEGFR2 (KDR) tyrosine kinase activity, demonstrating excellent selectivity versus other receptor tyrosine kinases. It inhibits VEGF-A-induced endothelial cell proliferation (IC50=8 nM) and VEGFR2 tyrosine phosphorylation, making it suitable for research applications.
  • Potent and selective VEGFR2 inhibitor
  • Anilinoquinazoline structure
  • Inhibits VEGF-A-induced endothelial cell proliferation
  • Inhibits VEGFR2 tyrosine phosphorylation
  • For research use only