Learn More
Medchemexpress LLC ZM39923 hydrochloride | 1021868-92-7 | MFCD04974534 | 99.8% | 367.91 g/mol | C23H26ClNO | 5MG
SDP
Catalog No. 5000236188
Encompass_Preferred
ZM39923 hydrochloride is a research-grade small molecule that acts primarily as a JAK3 inhibitor (pIC50 = 7.1) and also potently inhibits tissue transglutaminase (TGM2) (IC50 ≈ 10 nM). It displays weaker activity against EGFR and JAK1 and minimal activity versus Lck and CDK4. Supplied as the hydrochloride salt with high reported purity, it is intended for laboratory research use only.
  • JAK3 inhibitor with pIC50 7.1.
  • Potent tissue transglutaminase (TGM2) inhibition (IC50 ≈ 10 nM).
  • Hydrochloride salt form for improved stability and handling.
  • High reported purity (around 99.8%).
  • Available as solid and as a DMSO solution format.
  • Intended for research use only; not for diagnostic or therapeutic use.
Catalog No. 50-002-36188 Supplier Medchemexpress LLC Supplier No. HY125895MG
ZM39923 hydrochloride is a research-grade small molecule that acts primarily as a JAK3 inhibitor (pIC50 = 7.1) and also potently inhibits tissue transglutaminase (TGM2) (IC50 ≈ 10 nM). It displays weaker activity against EGFR and JAK1 and minimal activity versus Lck and CDK4. Supplied as the hydrochloride salt with high reported purity, it is intended for laboratory research use only.
  • JAK3 inhibitor with pIC50 7.1.
  • Potent tissue transglutaminase (TGM2) inhibition (IC50 ≈ 10 nM).
  • Hydrochloride salt form for improved stability and handling.
  • High reported purity (around 99.8%).
  • Available as solid and as a DMSO solution format.
  • Intended for research use only; not for diagnostic or therapeutic use.