Learn More
Aviva Systems Biology Corp ZMAT1 Antibody - middle region (ARP50721_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50721_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZMAT1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ZMAT1. Alternate names: KIAA1789, MGC176728ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-533