Learn More
Aviva Systems Biology Corp ZMAT2 Antibody - middle region (ARP41126_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41126_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZMAT2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human ZMAT2. Alternate names: FLJ31121, Snu23, hSNU23, Ptg-12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-284-913