Learn More
Aviva Systems Biology Corp ZMAT2 Antibody - N-terminal region (ARP41127_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41127_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZMAT2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ZMAT2. Alternate names: FLJ31121, Snu23, hSNU23, Ptg-12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-914