Learn More
ABclonal Technology ZMAT2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A144561000UL
Encompass_Preferred
Predicted to enable DNA binding activity and zinc ion binding activity. Involved in mRNA splicing, via spliceosome. Located in nucleus. Part of U2-type precatalytic spliceosome.
Catalog No. 50-001-08264