Learn More
Aviva Systems Biology Corp ZMIZ1 Antibody - N-terminal region (ARP39413_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39413_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZMIZ1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZMIZ1. Alternate names: FLJ13541, KIAA1224, MIZ, RAI17, Zimp10, hZIMP10, ZIMP10, TRAFIP10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57023