Learn More
Aviva Systems Biology Corp ZMIZ2 Antibody - middle region (ARP60635_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60635_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZMIZ2. It was validated on Western Blot. . Alternate names: DKFZp761I2123, KIAA1886, ZIMP7, hZIMP7, NET27, TRAFIP20ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-107